Learn More
Alkali Scientific CellPro™ Vacuum Filtration Flasks, 250mL, 0.45µm Membrane, PVDF, Individually Wrapped, 12/cs
SDP
Supplier:  Alkali Scientific VP25045
Encompass_Preferred
CellPro™ Filtration Flasks are available in filter medium PVDF 0.45µm. The screw-on funnel has a Lid, the filtration section has a graduated connection that fit multiple hoses, and the receiving bottle has indentations for easy gripping and a separate cap. Both the funnel and receiver feature engraved graduation marks to ensure accuracy, and are gamma sterilized, pyrogen free.
Catalog No. 50-233-5842