Learn More
Alkali Scientific Vacuum Filtration Flasks PES membrane, 500mL, 0.45um, Sterile
SDP
Supplier:  Alkali Scientific V50045
Encompass_Preferred
CellPro™ Filtration Flasks are available in filter medium PES 0.22µm and 0.45µm. The screw-on funnel has a lid, the filtration section has a graduated connection that fit multiple hoses, and the receiving bottle has indentations for easy gripping and a separate cap. Both the funnel and receiver feature engraved graduation marks to ensure accuracy, and are gamma sterilized, pyrogen free.
Catalog No. 50-233-5753