Learn More
Chemglass Life Sciences Adapter, Vacuum Filtration, 23mm ID Flange, #2 Pluro Stopper, 19/22 Lower Vacuum Assembly, Overall Height 110mm
SDP
Catalog No. CG105311
Encompass_Preferred
Adapter, Vacuum Filtration, 23mm ID Flange, #2 Pluro Stopper, 19/22 Lower Vacuum Assembly, Overall Height 110mm. Adapter used for reduced pressure filtration with plain stem Buchner funnels. Top is flared to accept the listed size pluro stopper. The hose connection has an O.D. of 10mm at the largest serration. ADAPTER IS NOT SUPPLIED WITH RUBBER PLURO STOPPER, See CG-1401.
Catalog No. CG105311 Supplier Chemglass Life Sciences Supplier No. CG105311
Adapter, Vacuum Filtration, 23mm ID Flange, #2 Pluro Stopper, 19/22 Lower Vacuum Assembly, Overall Height 110mm. Adapter used for reduced pressure filtration with plain stem Buchner funnels. Top is flared to accept the listed size pluro stopper. The hose connection has an O.D. of 10mm at the largest serration. ADAPTER IS NOT SUPPLIED WITH RUBBER PLURO STOPPER, See CG-1401.