Learn More
Alkali Scientific CellPro™ Vacuum Filtration Flasks, PVDF, 0.22µm Membrane, 150mL, Individually Wrapped, Sterile
SDP
Supplier:  Alkali Scientific VP15022
Encompass_Preferred
CellPro™ Filtration Flasks are available in filter medium PVDF 0.22µm. The screw-on funnel has a lid, the filtration section has a graduated connection that fit multiple hoses, and the receiving bottle has indentations for easy gripping and a separate cap. Both the funnel and receiver feature engraved graduation marks to ensure accuracy, and are gamma sterilized, pyrogen free.
Catalog No. 50-238-9042