Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Ground Joint Flasks for Vacuum Filtration

Catalog No. xx1614706 Shop All MilliporeSigma Products
Change view
Click to view available options
Volume (Metric):
250 mL
1 L
4 L
5 L
Type:
Conical Bottom Flask
Standard Bottom Flask
Tubing
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Type
XX1614706 5 L Conical Bottom Flask
XX1614705 1 L Conical Bottom Flask
XX1514705 1 L Standard Bottom Flask
XX7100004 250 mL Tubing
XX1014745 4 L Standard Bottom Flask
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. XX1614706 Supplier MilliporeSigma™ Supplier No. XX1614706
Only null left
Add to Cart
Add to Cart

Accessories for filter holders for sample preparation.

  • These accessory products are designed to replace parts contained in Millipore glass vacuum filtration kits or provide for an alternative configuration.
  • This item is an accessory part for the Millipore All-Glass Filter Holders Kits and Millipore MilliSolve Kit.
TRUSTED_SUSTAINABILITY

Specifications

Color Clear
Disposable Reusable
For Use With (Equipment) Millipore All-glass Filter Holders Kit, Millipore MilliSolve Kit
Material Borosilicate Glass
Shape Conical Bottom
Type Conical Bottom Flask
Volume (Metric) 5 L
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.