Learn More
Fischer Technical Company™ Vacuum Gauge and Fittings Kit for LAV Series
SCI_ED
Catalog No. S012380 Shop All Fischer Technical Company Products
Change view
Click to view available options
Pressure Range:
0 to 30 mmHg
Catalog No. Pressure Range
S012380 0 to 30 mmHg
1 options
Prop 65: Cancer and Reproductive Harm.
Catalog No. S012380 Supplier Fischer Technical Company™ Supplier No. HVG003
Add to Cart
Add to Cart
Prop 65: Cancer and Reproductive Harm.
Adds a gauge to your LAV Series Pump with this kit.
This kit comes with all of the fittings necessary to add a mounted gauge to your Fischer Technical Company™ LAV series High Vacuum Pump and includes the following:
  • 0 to 30 in. (2.5 in. dia.) Bourdon Tube Vacuum Gauge
  • Brass Tee
  • Hex Bushing
  • Hex Nipple
  • Vial of Vacuum Sealant
Specifications
Accuracy ±3% Full-scale
Pressure Range 0 to 30 mmHg
Display Type Analog
Product Type Vacuum/Pressure Gauge
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov