Learn More
PFEIFFER VACUUM INC Pfeiffer Vacuum Pirani Gauge, 80 °C Bakeout Temperature, DN 16 ISO-KF, 1E-4 to 1000 hPa
Catalog No. NC3978984
Encompass_Preferred
Pfeiffer Vacuum Pirani Gauge, 80 °C Bakeout Temperature, DN 16 ISO-KF, 1E-4 to 1000 hPa
Catalog No. NC3978984 Supplier PFEIFFER VACUUM INC Supplier No. 2000074754
Pfeiffer Vacuum Pirani Gauge, 80 °C Bakeout Temperature, DN 16 ISO-KF, 1E-4 to 1000 hPa