Learn More
PFEIFFER VACUUM INC Pfeiffer Vacuum Pirani/Capacitive Gauge, Metal Sealed, 80 °C, DN 16 ISO-KF
Catalog No. NC3978989
Encompass_Preferred
This Pirani/capacitive gauge is an active transmitter designed for precise vacuum measurement.
  • Metal sealed construction.
  • Maximum bakeout temperature of 80°C.
  • Equipped with a DN 16 ISO-KF connection flange.
  • Measures vacuum in the range of 5E-5 to 1.5E3 hPa.
  • Utilizes Pirani and Capacitive measuring methods.
  • Input voltage is 15 to 30 V DC.
  • Made with a stainless steel flange.
Catalog No. NC3978989 Supplier PFEIFFER VACUUM INC Supplier No. 2000006183
This Pirani/capacitive gauge is an active transmitter designed for precise vacuum measurement.
  • Metal sealed construction.
  • Maximum bakeout temperature of 80°C.
  • Equipped with a DN 16 ISO-KF connection flange.
  • Measures vacuum in the range of 5E-5 to 1.5E3 hPa.
  • Utilizes Pirani and Capacitive measuring methods.
  • Input voltage is 15 to 30 V DC.
  • Made with a stainless steel flange.