Learn More
PFEIFFER VACUUM INC Pfeiffer Vacuum Active Pirani/Cold Cathode Gauge, Metal Sealed, DN 40 CF-F
Catalog No. NC3978994
Encompass_Preferred
This gauge is designed for vacuum measurement of gases within a pressure range of 5 x 10-9 to 1000 hPa. It features two separate measurement systems, Pirani and Cold Cathode, combined into a single case to generate a uniform measurement signal. Optimized for high vacuum applications, it is rugged, dependable, insensitive to air ingress, and does not have an elastomer seal.
  • Flange size: DN 40 CF-F
  • Measurement range: 5 x 10-9 to 1000 hPa
  • Dual measurement systems (Pirani and Cold Cathode) within a single case
  • Corrosion-resistant and metal-sealed
  • Optimized for high vacuum applications
  • Rugged and dependable
  • Insensitive to air ingress
  • Without an elastomer seal
  • Maximum pressure applies to inert gases and temperatures below 55°C
Catalog No. NC3978994 Supplier PFEIFFER VACUUM INC Supplier No. 2000005754
This gauge is designed for vacuum measurement of gases within a pressure range of 5 x 10-9 to 1000 hPa. It features two separate measurement systems, Pirani and Cold Cathode, combined into a single case to generate a uniform measurement signal. Optimized for high vacuum applications, it is rugged, dependable, insensitive to air ingress, and does not have an elastomer seal.
  • Flange size: DN 40 CF-F
  • Measurement range: 5 x 10-9 to 1000 hPa
  • Dual measurement systems (Pirani and Cold Cathode) within a single case
  • Corrosion-resistant and metal-sealed
  • Optimized for high vacuum applications
  • Rugged and dependable
  • Insensitive to air ingress
  • Without an elastomer seal
  • Maximum pressure applies to inert gases and temperatures below 55°C