Learn More
PFEIFFER VACUUM INC Pfeiffer Vacuum Digital Pirani/Cold Cathode Gauge, DN 25 ISO-KF, 5E-9 to 1E3 hPa
Catalog No. NC3978992
Encompass_Preferred
This digital Pirani/cold cathode vacuum gauge offers precise measurement of gases, integrating both Pirani and cold cathode sensors into a single, compact unit for a wide measurement range suitable for various vacuum applications.
  • Combines Pirani and cold cathode sensors for comprehensive vacuum measurement.
  • Wide measurement range: 5 × 10-9 to 1000 hPa.
  • Digital RS-485, analog 0 to 10 V interfaces, and integrated display.
  • Cold cathode sensor features remote on/off control.
  • Corrosion-resistant materials: Tungsten, Stainless Steel, Nickel, and Molybdenum.
  • DN 25 ISO-KF connection flange with bakeout temperature up to 160 °C.
  • Includes 2 relay switching points; IP54 protection; low power consumption (max. 3.9 W).
Catalog No. NC3978992 Supplier PFEIFFER VACUUM INC Supplier No. 2000006447
This digital Pirani/cold cathode vacuum gauge offers precise measurement of gases, integrating both Pirani and cold cathode sensors into a single, compact unit for a wide measurement range suitable for various vacuum applications.
  • Combines Pirani and cold cathode sensors for comprehensive vacuum measurement.
  • Wide measurement range: 5 × 10-9 to 1000 hPa.
  • Digital RS-485, analog 0 to 10 V interfaces, and integrated display.
  • Cold cathode sensor features remote on/off control.
  • Corrosion-resistant materials: Tungsten, Stainless Steel, Nickel, and Molybdenum.
  • DN 25 ISO-KF connection flange with bakeout temperature up to 160 °C.
  • Includes 2 relay switching points; IP54 protection; low power consumption (max. 3.9 W).