Learn More
Chemglass Life Sciences Vacuum Gauge Trap
SDP
Catalog No. CG440801
Encompass_Preferred
Vacuum Gauge Trap. Trap provides protection of vacuum gauges from particulates which contaminate the mercury when the gauge is vented to the atmosphere. Supplied packed with glass wool. Hose connections have a 9mm OD at the middle serration.
Catalog No. CG440801 Supplier Chemglass Life Sciences Supplier No. CG440801
Vacuum Gauge Trap. Trap provides protection of vacuum gauges from particulates which contaminate the mercury when the gauge is vented to the atmosphere. Supplied packed with glass wool. Hose connections have a 9mm OD at the middle serration.