Learn More
MSC Guardair Vacuum Generating Engine, Diecast Aluminum, Generating Head, for PowerQUAD Vacuums
Catalog No. 502837277
Encompass
The PowerQUAD Vacuum Generating Engine is a rugged, diecast aluminum engine designed for use with PowerQUAD Vacuums. It functions as a generating head for drum-top vacuum heads, offering reliable performance for various applications.
  • Rugged, diecast aluminum vacuum-generating engine
  • Includes three color-coded, easy-to-change, nozzle sets
  • Nozzle sets are optimized for three progressive levels of performance and compressed air savings
  • Comes with vacuum engine, vacuum engine gasket, nozzle set, and installation hardware
Catalog No. 50-283-7277 Supplier MSC Supplier No. 64362486
The PowerQUAD Vacuum Generating Engine is a rugged, diecast aluminum engine designed for use with PowerQUAD Vacuums. It functions as a generating head for drum-top vacuum heads, offering reliable performance for various applications.
  • Rugged, diecast aluminum vacuum-generating engine
  • Includes three color-coded, easy-to-change, nozzle sets
  • Nozzle sets are optimized for three progressive levels of performance and compressed air savings
  • Comes with vacuum engine, vacuum engine gasket, nozzle set, and installation hardware