Learn More
MSC Guardair Hose Inlet Replacement Kit, 1.5 in ID Coupler, Nickel-Plated Steel
Catalog No. 502838911
Encompass
This vacuum hose inlet replacement kit is designed for 5 & 10 gallon drum covers. It features a nickel-plated steel inlet that fits a 1.5 inch ID vacuum hose coupler, and includes a gasket and hardware for installation.
  • Nickel-plated steel inlet fits 1.5 in ID vacuum hose coupler.
  • Includes gasket and hardware.
  • For use with 5 & 10 gallon vacuums.
  • Made in the USA.
  • OSHA compliant.
Catalog No. 50-283-8911 Supplier MSC Supplier No. 39466073
This vacuum hose inlet replacement kit is designed for 5 & 10 gallon drum covers. It features a nickel-plated steel inlet that fits a 1.5 inch ID vacuum hose coupler, and includes a gasket and hardware for installation.
  • Nickel-plated steel inlet fits 1.5 in ID vacuum hose coupler.
  • Includes gasket and hardware.
  • For use with 5 & 10 gallon vacuums.
  • Made in the USA.
  • OSHA compliant.