Learn More
Waters Corp Waters Vacuum Hose Assembly, 1 in., KF25 Adaptor, 1.5 m
Catalog No. 50791670 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This replacement vacuum hose assembly is designed to connect a rotary pump to mass spectrometers.
  • Features one KF25 adaptor and one open end.
  • Has a length of 1.5 m.
  • Typically used in a pair to connect a rotary pump to ACQUITY SQD, ACQUITY TQD, or 3100 Mass Spectrometers.
Catalog No. 50-791-670 Supplier Waters Corp Supplier No. 205000359
This replacement vacuum hose assembly is designed to connect a rotary pump to mass spectrometers.
  • Features one KF25 adaptor and one open end.
  • Has a length of 1.5 m.
  • Typically used in a pair to connect a rotary pump to ACQUITY SQD, ACQUITY TQD, or 3100 Mass Spectrometers.