Learn More
Mopec™ Vacuum Hydro Aspirator with Threaded Ends
SDP
Catalog No. 22445778 Shop All Mopec Products
Change view
Click to view available options
For Use With (Equipment):
Vacuum
Catalog No. For Use With (Equipment)
22-445-778 Vacuum
1 options
Catalog No. 22-445-778 Supplier Mopec™ Supplier No. BE013T
Add to Cart
Add to Cart
High vacuum maintains a 26 in. suction at 60 lb. of water pressure in this Vacuum Hydro Aspirator and can easily disassemble for cleaning.
  • Built-in vacuum breaker eliminates the possibility of back siphonage
  • Reverse flow allows for simple clearing of clogged suction line and provides fresh water spray 3 to 5 gallons per minute
  • Hose thread for easy attachment to most sinks
  • 40 to 60 psi
  • Standard 0.75 in. hose thread
  • Brass construction with high luster chrome finish
Specifications
For Use With (Equipment) Vacuum