Learn More
Synthware™ Vacuum or Inert Gas Manifold, Double, Front Left, Rear Right Connection
GREENER_CHOICESDP
Catalog No. 31501733 Shop All Kemtech Products
Change view
Click to view available options
No. of Ports:
4
5
Width (Metric):
450 mm
550 mm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Ports Width (Metric)
31-501-733 4 450 mm
31-501-734 5 550 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 31-501-733 Supplier Synthware™ Supplier No. M111004
Add to Cart
Add to Cart
General purpose double bank manifold for use with vacuum and inert gas.
Supplied with 0 to 4 mm high vacuum valves for grease-free operation. Both banks are made from 22 mm medium wall tubing and have all banks open. Ports are spaced 100 mm center to center and have an O.D. of 10 mm.
Specifications
Connection Type Front Left; Rear Right Hose Connection
Type Vacuum/Inert Gas Manifold
For Use With (Application) Organic Synthesis
Includes High Vacuum Valve Plugs
Material Borosilicate Glass
No. of Ports 4
Width (Metric) 450 mm