Learn More
Synthware™ Vacuum and Inert Gas Manifold with Hollow Glass Stopcocks
GREENER_CHOICESDP
Catalog No. 31501796 Shop All Kemtech Products
Change view
Click to view available options
Connection Type:
Front L/R Hose Connection, Rear Left Port
Front Left Hose Connection, Rear Right Port
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Connection Type
31-501-796 Front Left Hose Connection, Rear Right Port
31-501-797 Front L/R Hose Connection, Rear Left Port
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 31-501-796 Supplier Synthware™ Supplier No. M340004
Add to Cart
Add to Cart
No retention clip needed. Hollow stopcocks held in place by vacuum.
High vacuum hollow glass stopcocks precision ground with corresponding numbers on barrel and plug. Lower vacuum chambers on stopcock are evacuated with a separate hole in the plug in order to hold the plug in the stopcock barrels. -Hose connection : Back Beam- None, Front Beam- Left - 35/20 Socket on right side of back beam
Specifications
Connection Type Front Left Hose Connection, Rear Right Port
Type Double Manifold
For Use With (Application) Organic Synthesis
Includes 4 ports; Hollow glass stopcock
Material Borosilicate Glass
No. of Ports 4
Port Size (Metric) 35/20 Socket Joint