Learn More
Synthware™ Vacuum and Inert Gas Manifold with Hollow Glass Stopcocks, Front/Left, Rear/Left/Right Connection
GREENER_CHOICESDP
Catalog No. 31501757 Shop All Kemtech Products
Change view
Click to view available options
No. of Ports:
5
Catalog No. No. of Ports
31-501-757 5
1 options
Catalog No. 31-501-757 Supplier Synthware™ Supplier No. M441005
Add to Cart
Add to Cart
No retention clip needed. Hollow stopcocks held in place by vacuum.
High vacuum hollow glass stopcocks precision ground with corresponding numbers on barrel and plug. Lower vacuum chambers on stopcock are evacuated with a separate hole in the plug in order to hold the plug in the stopcock barrels. -Hose connection : Back Beam-Both Ends, Front Beam-Left Only
Specifications
Description Front Left; Rear Left and Right Hose Connection
Type Vacuum/Inert Gas Manifold
For Use With (Application) Organic Synthesis
Includes Hollow Glass Stopcocks
Material Borosilicate Glass
No. of Ports 5
Width (Metric) 450 mm