Learn More
Synthware™ Vacuum and Inert Gas Manifold with Hollow Glass Stopcocks, Front Left/Right, Rear Right Spcherical Connection
GREENER_CHOICESDP
Catalog No. 31501795 Shop All Kemtech Products
Change view
Click to view available options
No. of Ports:
4
5
Width (Metric):
365 mm
450 mm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Ports Width (Metric)
31-501-795 5 450 mm
31-501-794 4 365 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 31-501-795 Supplier Synthware™ Supplier No. M270005
Add to Cart
Add to Cart
No retention clip needed. Hollow stopcocks held in place by vacuum.
High vacuum hollow glass stopcocks precision ground with corresponding numbers on barrel and plug. Lower vacuum chambers on stopcock are evacuated with a separate hole in the plug in order to hold the plug in the stopcock barrels. -Hose connection : Back Beam- None, Front Beam- None - 35/20 Socket on right side of back beam and both sides of front beam.
Specifications
Description Front Left and Right; Rear Right Spherical Connection
Type Spherical Joint Double Manifold
For Use With (Application) Organic Synthesis
Includes Hollow Glass Stopcocks
Material Borosilicate Glass
No. of Ports 5
Width (Metric) 450 mm