Learn More
Ace Glass, Inc. VACUUM LINE C VACUUM LINE C
SDP
Catalog No. 50975479
Encompass_Preferred
Supplemental Vacuum Line with McLeod Gauge, for the ACE-Burlitch system. The ACE-Burlitch inert atmosphere system is designed for maximum convenience and versatility in the handling of air-sensitive compounds in conjunction with ACE no-air labware, or suitiably adapted conventional glassware.
Catalog No. 50-975-479 Supplier Ace Glass, Inc. Supplier No. 781887
Add to Cart
Add to Cart
Supplemental Vacuum Line with McLeod Gauge, for the ACE-Burlitch system. The ACE-Burlitch inert atmosphere system is designed for maximum convenience and versatility in the handling of air-sensitive compounds in conjunction with ACE no-air labware, or suitiably adapted conventional glassware.