Learn More
Chemglass Life Sciences Vacuum/Inert Manifold, 4 Ports, 500mm Overall Width
SDP
Supplier:  Chemglass Life Sciences CG443802
Encompass_Preferred
Vacuum/Inert Manifold, 4 Ports, 500mm Overall Width. General purpose double bank vacuum manifold with 3, 4, or 5 ports. Both tubes are 3/4 OD with serrated hose connection, having the smallest serration 13.5mm and the largest serration 15.5mm, located on the upper right and lower left banks. Ports are 0-4mm Chem-Vac™ Chem-Cap valves with serrated hose connection having the smallest serration 9mm and the largest serration 10mm. Ports are spaced 100mm center to center. Manifold has rod braces sealed between each bank located in-line with the valves which are sealed at a 45 angle to allow for easier access.
Catalog No. CG443802