Learn More
Synthware™ Vacuum Manifold, Double Bank
GREENER_CHOICESDP
Catalog No. 31501806 Shop All Kemtech Products
Change view
Click to view available options
Material:
Borosilicate Glass
Catalog No. Material
31-501-806 Borosilicate Glass
1 options
Catalog No. 31-501-806 Supplier Synthware™ Supplier No. M330804
Add to Cart
Add to Cart
Vacuum Manifold, Double Bank with forward facing high vacuum valves allow easy operation of manifold inside of a fume hood. Main parts are spaced 100 mm center to center. Adapter components or stoppers are available and have to be ordered separately.
  • 4 Ports
  • 0 to 8 mm High Vacuum Valves
Specifications
Connection Type Front Left and Right, Rear Left and Right Spherical Connection
Description Front Left and Right; Rear Left and Right Spherical Connection
Type Double Manifold
For Use With (Application) Organic Synthesis
Includes #15 O-ring Joints
Material Borosilicate Glass