Learn More
Electron Microscopy Sciences Vacuum Vacuum Mounting Blocks, 2.125"
SDP
Catalog No. 50980967
Encompass
Vacuum Mounting Bocks are for use with the Models 155DV, 155D, and 155V lapping and polishing fixture. Made of stainless steel. 2" (50mm) Diameter Vacuum Pattern.
Diameter: 2.125" (54mm)
Use: 155D, 155V
Catalog No. 50-980-967 Supplier Electron Microscopy Sciences Supplier No. 5015112
Vacuum Mounting Bocks are for use with the Models 155DV, 155D, and 155V lapping and polishing fixture. Made of stainless steel. 2" (50mm) Diameter Vacuum Pattern.
Diameter: 2.125" (54mm)
Use: 155D, 155V