Learn More
Electron Microscopy Sciences Vacuum Vacuum Mounting Blocks, 2.186"
SDP
Catalog No. 50980966
Encompass
Vacuum Mounting Bocks are for use with the Models 155DV, 155D, and 155V lapping and polishing fixture. Made of stainless steel. Blank for Vacuum Pattern.
Diameter: 2.186" (55.5mm)
Use: 155D, 155V, 155DV
Catalog No. 50-980-966 Supplier Electron Microscopy Sciences Supplier No. 5015111
Vacuum Mounting Bocks are for use with the Models 155DV, 155D, and 155V lapping and polishing fixture. Made of stainless steel. Blank for Vacuum Pattern.
Diameter: 2.186" (55.5mm)
Use: 155D, 155V, 155DV