Learn More
Electron Microscopy Sciences Vacuum Oven Model 3606, 240 V, 8"W x 8"H x 12"D chamber, 0.44 cu. ft (12.5L) Capacity
SDP
Catalog No. 5031306
Encompass_Preferred
Top-mounted independent evacuation and venting vacuum fittings.
Chamber: 8"W x 8"H x 12"D
Capacity: 0.44 cu. ft (12.5L)
Thermometer: Dial
Catalog No. 50-313-06 Supplier Electron Microscopy Sciences Supplier No. 6323420
Top-mounted independent evacuation and venting vacuum fittings.
Chamber: 8"W x 8"H x 12"D
Capacity: 0.44 cu. ft (12.5L)
Thermometer: Dial