Learn More
Electron Microscopy Sciences Vacuum Oven Model 3618, 120 V, 14"W x 14"H x 20"D chamber, 2.3 cu. ft (65 L) Capacity
SDP
Catalog No. 5031309
Encompass_Preferred
Multipurpose vacuum oven with 220 deree C maximum temperature conserves chamber space for drying, curing, vacuum embedding, and plating applications. Silicone door gasket and a positive latch door maintains the seal for all vacuum levels. Exclusive Polycarbonate safety shield protects door glass. Two removable aluminum shelves including standard. Easy-to-clean #304 staniless steel chamber interior. Powder coated heavy guage steel exterior construction.
Chamber: 14"W x 14"H x 20"D
Capacity: 2.3 cu. ft (65 L)
Thermometer: Dial
Catalog No. 50-313-09 Supplier Electron Microscopy Sciences Supplier No. 6323710
Add to Cart
Add to Cart
Request a Quote
Multipurpose vacuum oven with 220 deree C maximum temperature conserves chamber space for drying, curing, vacuum embedding, and plating applications. Silicone door gasket and a positive latch door maintains the seal for all vacuum levels. Exclusive Polycarbonate safety shield protects door glass. Two removable aluminum shelves including standard. Easy-to-clean #304 staniless steel chamber interior. Powder coated heavy guage steel exterior construction.
Chamber: 14"W x 14"H x 20"D
Capacity: 2.3 cu. ft (65 L)
Thermometer: Dial