Learn More
Electron Microscopy Sciences Vacuum Pick-Up System, 115 V
SDP
Catalog No. 50949640
Encompass_Preferred
VaccuumPick Up handles delicate objects in the laboratory. The system avoids contamination of parts and performs functions that tweezers does such as sorting, picking up, holding, carrying, and transferring. It is ideal for handling cover slips without the use of forceps.
Light weight, completely assembled and ready for immediate operation as soon as the proper tip is selected and installed. The vacuum generator has an adjustable vacuum pressure control from 1" Hg to 15" Hg.
Vacuum pick-up system includes: vacuum generator, aluminum vacuum pen, five vacuum tips, set of seven rubber vacuum cups (size ranging: 9⁄16", 1/2", 7⁄16", 3⁄8", 5⁄16", 1⁄4" and 3⁄16"), an in-line filter, and 4 ft (122cm) of vacuum tubing.
Catalog No. 50-949-640 Supplier Electron Microscopy Sciences Supplier No. 71894
VaccuumPick Up handles delicate objects in the laboratory. The system avoids contamination of parts and performs functions that tweezers does such as sorting, picking up, holding, carrying, and transferring. It is ideal for handling cover slips without the use of forceps.
Light weight, completely assembled and ready for immediate operation as soon as the proper tip is selected and installed. The vacuum generator has an adjustable vacuum pressure control from 1" Hg to 15" Hg.
Vacuum pick-up system includes: vacuum generator, aluminum vacuum pen, five vacuum tips, set of seven rubber vacuum cups (size ranging: 9⁄16", 1/2", 7⁄16", 3⁄8", 5⁄16", 1⁄4" and 3⁄16"), an in-line filter, and 4 ft (122cm) of vacuum tubing.