Learn More
World Precision Instrument Epoxy Coated Stainless Steel Tweezers
SDP
Catalog No. NC2225222
Encompass
  • The soft feel and non-slip grip makes this kit ideal for many applications. The tweezers are non-magnetic and non-reflective with an epoxy coating.
This item is not returnable. View return policy
Catalog No. NC2225222 Supplier World Precision Instrument Supplier No. WPK570455
  • The soft feel and non-slip grip makes this kit ideal for many applications. The tweezers are non-magnetic and non-reflective with an epoxy coating.