Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fischer Technical Company™ Vacuum Pressure Aspirator Bulb
SCI_ED

Catalog No. S2335018 Shop All Fischer Technical Company Products
Change view
Click to view available options
Material:
Natural Rubber
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
S2335018 Natural Rubber
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S2335018 Supplier Fischer Technical Company™ Supplier No. VPBULB
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For use with Gas Analysis Applications.

Our PVC deluxe Vacuum Pressure Bulb is ideal for gas aspirating duties.

  • Natural rubber bulb
  • Red valves are interchangeable and can be used as both a pressure and vacuum valve
  • 111 mm length
  • 57 mm diameter

Specifications

Adjustable Tip Spacing No
Autoclavable Not Autoclavable
Color Red
Disposable Reusable
Grip Type Ergonomic
Material Natural Rubber
Model VPBULB
Number of Channels 1
Sterility Non-sterile
Type Vacuum Pressure Aspirator Bulb
Quantity 1 Pk.
Length (Metric) 13 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.