Learn More
Electron Microscopy Sciences Vacuum Pressure Indicator, Clear
SDP
Catalog No. 501899538
Encompass_Preferred
Color: Clear. This Vacuum-Pressure Indicator fits between the desiccator dome and the desiccator stopcock. Its slips easily onto the tapered outlet of the desiccator dome and into the standard desiccator stopcock without requiring any tools.. The thermo-plastic elastometer collapses when there is negative pressure vacuum in the desiccator and expands under ambient atmospheric pressure.
Catalog No. 50-189-9538 Supplier Electron Microscopy Sciences Supplier No. 7124201
Color: Clear. This Vacuum-Pressure Indicator fits between the desiccator dome and the desiccator stopcock. Its slips easily onto the tapered outlet of the desiccator dome and into the standard desiccator stopcock without requiring any tools.. The thermo-plastic elastometer collapses when there is negative pressure vacuum in the desiccator and expands under ambient atmospheric pressure.