Learn More
Gentech Scientific Inc Ulvac MSD Vacuum Pump (GHD-031)
SDP
Catalog No. 501897505
Encompass_Preferred
• Motor - Single phase, (50/60 Hz) 100-120V ,100W , 2 poles, capacitor run • Inlet, outlet pipe diameter - KF-16 • Ultimate pressure - 0.67 Pa • 1 Year Warranty GenTech Scientific offers installation, and extended warranty options. Please contact 585-492-1068 with questions. (We also offer a 220V)
Catalog No. 50-189-7505 Supplier Gentech Scientific Inc Supplier No. GTS 10630
• Motor - Single phase, (50/60 Hz) 100-120V ,100W , 2 poles, capacitor run • Inlet, outlet pipe diameter - KF-16 • Ultimate pressure - 0.67 Pa • 1 Year Warranty GenTech Scientific offers installation, and extended warranty options. Please contact 585-492-1068 with questions. (We also offer a 220V)