Learn More
Grainger VACUUM PUMP 115/208-230V AC
Catalog No. NC2160230
Encompass_Preferred
acuum Pump: 1/4 hp, 1 Phase, 115/208-230V AC, 4.2 cfm, 26 in Hg Max
This item is not returnable. View return policy
Catalog No. NC2160230 Supplier Grainger Supplier No. 3KYY4
acuum Pump: 1/4 hp, 1 Phase, 115/208-230V AC, 4.2 cfm, 26 in Hg Max