Learn More
Waters Corp Waters Corporation Vacuum Pump, 220V/240V, 50Hz
Catalog No. 502039915 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Vacuum pumps are essential lab equipment for chromatographic analysis and system design, offering compatibility with various analyses for universal laboratory use. They feature a single gauge with pressure controlled by a relief valve on the manifold. These pumps are a critical element of the Vacuum Manifold, especially for SPE cartridges, processing up to twenty samples simultaneously.
  • Essential for chromatographic analysis and system design.
  • Compatible with various analyses for universal lab use.
  • Features a single gauge with pressure controlled by a relief valve.
  • Critical for the Vacuum Manifold, especially with SPE cartridges.
  • Processes up to twenty samples simultaneously with the Vacuum Manifold.
  • Operates at 220V/240V, 50Hz.
Catalog No. 50-203-9915 Supplier Waters Corp Supplier No. 725000604
Vacuum pumps are essential lab equipment for chromatographic analysis and system design, offering compatibility with various analyses for universal laboratory use. They feature a single gauge with pressure controlled by a relief valve on the manifold. These pumps are a critical element of the Vacuum Manifold, especially for SPE cartridges, processing up to twenty samples simultaneously.
  • Essential for chromatographic analysis and system design.
  • Compatible with various analyses for universal lab use.
  • Features a single gauge with pressure controlled by a relief valve.
  • Critical for the Vacuum Manifold, especially with SPE cartridges.
  • Processes up to twenty samples simultaneously with the Vacuum Manifold.
  • Operates at 220V/240V, 50Hz.