Learn More
PFEIFFER VACUUM INC PFEIFFER VACUUM Connection Kit, DN 25 ISO-KF, Corrugated Hose, 1 m
Catalog No. NC3978997
Encompass_Preferred
This connection kit is designed for mounting pumps and is suitable for high vacuum (HV) applications. It includes a 1 m corrugated hose, two seals, and two clamping rings, and features a DN 25 ISO-KF connection flange.
  • Connection kit for mounting of pumps.
  • Contains a corrugated hose (1 m), two seals, and two clamping rings.
  • Suitable for HV applications.
  • Connection flange: DN 25 ISO-KF.
  • A: 1000 mm.
  • B: 24 mm.
  • C: 33 mm.
Catalog No. NC3978997 Supplier PFEIFFER VACUUM INC Supplier No. 2000048029
This connection kit is designed for mounting pumps and is suitable for high vacuum (HV) applications. It includes a 1 m corrugated hose, two seals, and two clamping rings, and features a DN 25 ISO-KF connection flange.
  • Connection kit for mounting of pumps.
  • Contains a corrugated hose (1 m), two seals, and two clamping rings.
  • Suitable for HV applications.
  • Connection flange: DN 25 ISO-KF.
  • A: 1000 mm.
  • B: 24 mm.
  • C: 33 mm.