Learn More
PFEIFFER VACUUM INC Vacuum Pump Accessories, Pfeiffer Vacuum Inc., DN 40 ISO-KF 1 Meter Connection Kit
Catalog No. NC3978999
Encompass_Preferred
Vacuum Pump Accessories, Pfeiffer Vacuum Inc., DN 40 ISO-KF 1 Meter Connection Kit, Tubing Material: Stainless Steel, Accessories Included: Tubing + Clamps + O-Rings
Catalog No. NC3978999 Supplier PFEIFFER VACUUM INC Supplier No. 2000048030
Vacuum Pump Accessories, Pfeiffer Vacuum Inc., DN 40 ISO-KF 1 Meter Connection Kit, Tubing Material: Stainless Steel, Accessories Included: Tubing + Clamps + O-Rings