Learn More
Thermo Scientific Degasser Chamber System for Analyzers
Catalog No. 501347658 Shop All Thermo Scientific Products
Encompass
A degassing chamber removes dissolved gases from analytical liquid samples by drawing a vacuum to lower pressure and allow gases to come out of solution, improving accuracy for liquid chromatography.
Catalog No. 50-134-7658 Supplier Thermo Scientific Supplier No. 6030.9289
A degassing chamber removes dissolved gases from analytical liquid samples by drawing a vacuum to lower pressure and allow gases to come out of solution, improving accuracy for liquid chromatography.