Learn More
BINDER™ Vacuum Pump Connection Kit for VAP 1 and VAP 2
SDP
Catalog No. 08111961 Shop All BINDER Inc. Products
Change view
Click to view available options
Chamber Material:
Stainless Steel
Catalog No. Chamber Material
08-111-961 Stainless Steel
1 options
Catalog No. 08-111-961 Supplier BINDER™ Supplier No. 80121995
The Vacuum Pump Connection Kit contains all necessary connection parts for connecting Binder vacuum pump VAP 1 and VAP 2 with vacuum drying chambers.
Includes:
  • Aluminum clamping ring
  • Universal centering ring
  • Small flange with hose nozzle
  • 2.5 m rubber hose
  • 2 hose clamps
Specifications
Chamber Material Stainless Steel
Description Vacuum pump connection kit - for vacuum pump VAP1 and VAP2; consisting of aluminum clamping ring, universal centering ring, small flange with hose nozzle, (2.5 m) rubber hose and 2 hose clamps
For Use With (Equipment) BINDER Chambers Series VD
Type Vacuum Pump Connection Kit
Plug Type EU