Learn More
Research Products International Corp Vacuum Pump, High Capacity
SDP
Catalog No. 501976982
Encompass
This direct drive vacuum pump delivers the best performance at a great value. Two stage pump produces one micron ultimate vacuum at laboratory conditions. Direct drive system combined with a rotary sliding vane provides 3 CFM and operates quietly.
Additional features include a gas ballast for moisture protection and built in trap to prevent back flow of oil. Compact size allows pump to be placed at a convenient location.
Catalog No. 50-197-6982 Supplier Research Products International Corp Supplier No. 247830
This direct drive vacuum pump delivers the best performance at a great value. Two stage pump produces one micron ultimate vacuum at laboratory conditions. Direct drive system combined with a rotary sliding vane provides 3 CFM and operates quietly.
Additional features include a gas ballast for moisture protection and built in trap to prevent back flow of oil. Compact size allows pump to be placed at a convenient location.