Learn More
Chemglass Life Sciences BrandTech RC6 Chemistry-HYBRID Vacuum Pump, 120V, 50-60Hz
SDP
Catalog No. 501531768
Encompass_Preferred
BrandTech RC6 Chemistry-HYBRID Vacuum Pump, 120V, 50-60Hz. The BrandTech Hybrid pump combines a two stage rotary vane direct drive pump backed with a diaphragm pump. This design reduces the outlet pressure, reducing internal condensation in the oil section, and continuously cleans the oil. This also enhances pump lifetime.
Catalog No. 50-153-1768 Supplier Chemglass Life Sciences Supplier No. CG480301
Add to Cart
Add to Cart
Request a Quote
BrandTech RC6 Chemistry-HYBRID Vacuum Pump, 120V, 50-60Hz. The BrandTech Hybrid pump combines a two stage rotary vane direct drive pump backed with a diaphragm pump. This design reduces the outlet pressure, reducing internal condensation in the oil section, and continuously cleans the oil. This also enhances pump lifetime.