Learn More
Memmert Noise insulated Vacuum pump module with built in pump for VO49
SDP
Catalog No. 502254307
Encompass_Preferred
The corresponding pump module by Memmert - for use with the Memmert Vacuum Oven - VO 49. This noise-insulated, variable-speed, vacuum pump with stainless steel housing includes Vacuum Hose Connection Kit, Signal Cable for Pump Control, and Power cord.
This item is not returnable. View return policy
Catalog No. 50-225-4307 Supplier Memmert Supplier No. PMP49 230V
Add to Cart
Add to Cart
Request a Quote
The corresponding pump module by Memmert - for use with the Memmert Vacuum Oven - VO 49. This noise-insulated, variable-speed, vacuum pump with stainless steel housing includes Vacuum Hose Connection Kit, Signal Cable for Pump Control, and Power cord.