Learn More
Grainger Elmo Rietschle Multi-Lube 100 Oil, ISO 100, 1 Quart
Catalog No. 502794178
Encompass_Preferred
Multi-Lube 100 Oil is a high-quality lubricant specifically engineered for use with vacuum pumps and compressors. This OEM-certified oil ensures optimal performance and reliability for demanding industrial applications.
  • ISO viscosity grade 100
  • Optimized for use in vacuum pumps and compressors
  • Genuine OEM lubricant ensures equipment longevity
  • Supplied in a 1 qt. container
Catalog No. 50-279-4178 Supplier Grainger Supplier No. 1ARZ3
Multi-Lube 100 Oil is a high-quality lubricant specifically engineered for use with vacuum pumps and compressors. This OEM-certified oil ensures optimal performance and reliability for demanding industrial applications.
  • ISO viscosity grade 100
  • Optimized for use in vacuum pumps and compressors
  • Genuine OEM lubricant ensures equipment longevity
  • Supplied in a 1 qt. container