Learn More
PFEIFFER VACUUM INC Pfeiffer Vacuum Mineral Oil, Rotary Vane Pump, 20 L, Oxidation-Resistant
Catalog No. NC3979032
Encompass_Preferred
This high-quality mineral oil is designed for rotary vane vacuum pumps. It is oxidation-resistant and suitable for applications with high operating temperatures up to 95 °C.
  • Suitable for standard applications to pump down air, inert gases, and noble gases.
  • Achieves ultimate pressure of < 1 · 10-3 hPa.
  • Features extremely low vapor pressure.
  • Contains highly refined mineral oils and additives.
Catalog No. NC3979032 Supplier PFEIFFER VACUUM INC Supplier No. 2000025860
This high-quality mineral oil is designed for rotary vane vacuum pumps. It is oxidation-resistant and suitable for applications with high operating temperatures up to 95 °C.
  • Suitable for standard applications to pump down air, inert gases, and noble gases.
  • Achieves ultimate pressure of < 1 · 10-3 hPa.
  • Features extremely low vapor pressure.
  • Contains highly refined mineral oils and additives.