Learn More
PFEIFFER VACUUM INC Pfeiffer Vacuum P3 Mineral Oil, Rotary Vane Pump, 5 L, Hydrocarbon, Oxidation-Resistant
Catalog No. NC3979041
Encompass_Preferred
P3 mineral oil is a high-quality mineral oil designed for standard applications in rotary vane pumps. It is suitable for pumping air, inert gases, and noble gases, and can withstand operating temperatures up to 95 °C. This oil features extremely low vapor pressure and is oxidation-resistant, contributing to an achievable ultimate pressure of less than 1 · 10-3 hPa. The operating fluid amount is 5 L.
  • High-quality mineral oil for standard applications
  • Used for pumping air, inert gases, and noble gases
  • Suitable for high operating temperatures up to a maximum of 95 °C
  • Extremely low vapor pressure (1 · 10-3 hPa)
  • Oxidation-resistant
  • Designed for use with rotary vane pumps
  • Volume: 5 L
  • Achievable final pressure of less than 1 · 10-3 hPa
Catalog No. NC3979041 Supplier PFEIFFER VACUUM INC Supplier No. 2000025859
P3 mineral oil is a high-quality mineral oil designed for standard applications in rotary vane pumps. It is suitable for pumping air, inert gases, and noble gases, and can withstand operating temperatures up to 95 °C. This oil features extremely low vapor pressure and is oxidation-resistant, contributing to an achievable ultimate pressure of less than 1 · 10-3 hPa. The operating fluid amount is 5 L.
  • High-quality mineral oil for standard applications
  • Used for pumping air, inert gases, and noble gases
  • Suitable for high operating temperatures up to a maximum of 95 °C
  • Extremely low vapor pressure (1 · 10-3 hPa)
  • Oxidation-resistant
  • Designed for use with rotary vane pumps
  • Volume: 5 L
  • Achievable final pressure of less than 1 · 10-3 hPa