Learn More
Chemglass Life Sciences BrandTech ME4C NT Oil-Free PTFE Diaphragm Vacuum Pump, 120V, 73L/min, 23lbs., 10.5"(L)x9.5"(W)x7"(H)
SDP
Catalog No. 501531770
Encompass_Preferred
BrandTech ME4C NT Oil-Free PTFE Diaphragm Vacuum Pump, 120V, 73L/min, 23lbs., 10.5"(L)x9.5"(W)x7"(H). A popular vacuum pump suitable for many laboratory tasks, vacuum filtration, degassing, solid phase extraction and many other tasks. Ultimate Vacuum: 52 torr (70 mbar). Free Air Capacity: 73 lpm (2.6 cfm). Whisper quiet, performs more work, and has excellent corrosion resistance.
Catalog No. 50-153-1770 Supplier Chemglass Life Sciences Supplier No. CG480602
BrandTech ME4C NT Oil-Free PTFE Diaphragm Vacuum Pump, 120V, 73L/min, 23lbs., 10.5"(L)x9.5"(W)x7"(H). A popular vacuum pump suitable for many laboratory tasks, vacuum filtration, degassing, solid phase extraction and many other tasks. Ultimate Vacuum: 52 torr (70 mbar). Free Air Capacity: 73 lpm (2.6 cfm). Whisper quiet, performs more work, and has excellent corrosion resistance.