Learn More
Chemglass Life Sciences BrandTech MZ2C NT Oil-Free PTFE Diaphragm Vacuum Pump, 120V, 38L/min, 23lbs., 10.5"(L)x9.5"(W)x7"(H)
SDP
Catalog No. 501531771
Encompass_Preferred
BrandTech MZ2C NT Oil-Free PTFE Diaphragm Vacuum Pump, 120V, 38L/min, 23lbs., 10.5"(L)x9.5"(W)x7"(H). A popular vacuum pump suitable for many laboratory tasks, including rotary evaporation, centrifugal concentration, gel drying, vacuum filtration, solid phase extraction and many other tasks. Ultimate Vacuum: 5 torr (7 mbar). Free Air Capacity: 38 lpm (1.4 cfm). Whisper quiet, performs more work, and has excellent corrosion resistance.
Catalog No. 50-153-1771 Supplier Chemglass Life Sciences Supplier No. CG480603
BrandTech MZ2C NT Oil-Free PTFE Diaphragm Vacuum Pump, 120V, 38L/min, 23lbs., 10.5"(L)x9.5"(W)x7"(H). A popular vacuum pump suitable for many laboratory tasks, including rotary evaporation, centrifugal concentration, gel drying, vacuum filtration, solid phase extraction and many other tasks. Ultimate Vacuum: 5 torr (7 mbar). Free Air Capacity: 38 lpm (1.4 cfm). Whisper quiet, performs more work, and has excellent corrosion resistance.