Learn More
Chemglass Life Sciences BrandTech MD1C Oil-Free PTFE Diaphragm Vacuum Pump, 25L/min, 1.5 Torr Dual-Voltage, (220-230V, 50-60Hz / 100-115V, 50-60Hz / 120V, 60Hz)
SDP
Catalog No. 501531772
Encompass_Preferred
BrandTech MD1C Oil-Free PTFE Diaphraagm Vacuum Pump, 120V, 60Hz, 25L/min, 15lbs, 12.4"(L) x 5.6"(W) x 8.8"(H). Optimize rotary evaporation and other laboratory applications with theVACUUBRAND MD1C oil-free diaphragm vacuum pump. Compact and whisper-quiet, this pump is suitable for most benchtop rotary evaporators, and powerful enough for excellent results with high boiling point solvents. Ultimate Vacuum: 2 mbar, Free Air Capacity: 0.88 cfm. Significantly Quieter Than Rotary Vane Pumps, Perform More Work, Excellent Corrosion Resistance.
Catalog No. 50-153-1772 Supplier Chemglass Life Sciences Supplier No. CG480604
BrandTech MD1C Oil-Free PTFE Diaphraagm Vacuum Pump, 120V, 60Hz, 25L/min, 15lbs, 12.4"(L) x 5.6"(W) x 8.8"(H). Optimize rotary evaporation and other laboratory applications with theVACUUBRAND MD1C oil-free diaphragm vacuum pump. Compact and whisper-quiet, this pump is suitable for most benchtop rotary evaporators, and powerful enough for excellent results with high boiling point solvents. Ultimate Vacuum: 2 mbar, Free Air Capacity: 0.88 cfm. Significantly Quieter Than Rotary Vane Pumps, Perform More Work, Excellent Corrosion Resistance.