Learn More
Chemglass Life Sciences BrandTech ME2C NT Oil-Free One Head Chemistry PTFE Diaphragm Vacuum Pump, Single Stage, 120V, 38L/min, 16lbs, 10"(L) x 6.5"(W) x 7.5"(H)
SDP
Catalog No. 501531769
Encompass_Preferred
BrandTech ME2C NT Oil-Free One Head Chemistry PTFE Diaphragm Vacuum Pump, Single Stage, 120V, 38L/min, 16lbs, 10"(L) x 6.5"(W) x 7.5"(H). A vacuum pump excellent for many laboratory fluid movement applications including vacuum filtration and solid phase extraction, as well as degassing, and evaporation of volatile solvents. Ultimate vacuum: 52Torr (70 mbar), Free air capacity: 38lpm (1.4 cfm). Whisper quiet, performs more work, and has excellent corrosion resistance.
Catalog No. 50-153-1769 Supplier Chemglass Life Sciences Supplier No. CG480601
BrandTech ME2C NT Oil-Free One Head Chemistry PTFE Diaphragm Vacuum Pump, Single Stage, 120V, 38L/min, 16lbs, 10"(L) x 6.5"(W) x 7.5"(H). A vacuum pump excellent for many laboratory fluid movement applications including vacuum filtration and solid phase extraction, as well as degassing, and evaporation of volatile solvents. Ultimate vacuum: 52Torr (70 mbar), Free air capacity: 38lpm (1.4 cfm). Whisper quiet, performs more work, and has excellent corrosion resistance.