Learn More
Hoefer, Inc VP200 Vacuum Pump, Universal VAC. Companion pump to GD2000 Vacuum Gel Dryer
SDP
Catalog No. NC3790231
Encompass_Preferred
"The VP200 Vacuum Pump is ideal for use with the Hoefer GD2000 Vacuum Gel Dryer. It resists water vapor, acids, and gel staining solution vapors. Two vapor trap flasks at the inlet and outlet capture condensate and are secured with single clamps for easy cleaning. The pump is robust, user-friendly, and low-maintenance.Specifications:• Max Pump Speed: 1.2 cfm (120V 60Hz), 1.7/2.0 m³/h (230V 50/60Hz)• Ultimate Pressure: 9 mbar (20 mbar w/ ballast)• Max Outlet Pressure: 2 bar• Motor: 180W, overload cutout• Current Draw: 2.9A (120V 60Hz), 1.5/1.45A (230V ~50/60Hz)• Degree of Protection IEC 529: IP54• Temp: 0–40°C (operating), –10–60°C (storage)• Operating Relative Humidity: 30–85%, non-condensing• Sound Pressure Level: 41 dBA at 1 m• Rated Motor Speed at 50/60 Hz: 1500/1800 rpm• Inlet/Outlet Port Hose Nozzle Tubing Size: 10 mm ID, 1/8–27 NPT• Size: 31 x 29.5 x 24 cm• Weight: 11.7 kg"
Catalog No. NC3790231 Supplier Hoefer, Inc Supplier No. VP200
"The VP200 Vacuum Pump is ideal for use with the Hoefer GD2000 Vacuum Gel Dryer. It resists water vapor, acids, and gel staining solution vapors. Two vapor trap flasks at the inlet and outlet capture condensate and are secured with single clamps for easy cleaning. The pump is robust, user-friendly, and low-maintenance.Specifications:• Max Pump Speed: 1.2 cfm (120V 60Hz), 1.7/2.0 m³/h (230V 50/60Hz)• Ultimate Pressure: 9 mbar (20 mbar w/ ballast)• Max Outlet Pressure: 2 bar• Motor: 180W, overload cutout• Current Draw: 2.9A (120V 60Hz), 1.5/1.45A (230V ~50/60Hz)• Degree of Protection IEC 529: IP54• Temp: 0–40°C (operating), –10–60°C (storage)• Operating Relative Humidity: 30–85%, non-condensing• Sound Pressure Level: 41 dBA at 1 m• Rated Motor Speed at 50/60 Hz: 1500/1800 rpm• Inlet/Outlet Port Hose Nozzle Tubing Size: 10 mm ID, 1/8–27 NPT• Size: 31 x 29.5 x 24 cm• Weight: 11.7 kg"