Learn More
Cole-Parmer VACUUM PUMP, WITH GAUGE
Catalog No. NC1824548 Shop All Cole-Parmer Products
Encompass_Preferred
High-Capacity Vacuum Pump, with Gauge; 1.15 cfm, 29.0"Hg, 115V AC
Catalog No. NC1824548 Supplier Cole-Parmer Supplier No. 0706111
High-Capacity Vacuum Pump, with Gauge; 1.15 cfm, 29.0"Hg, 115V AC