Learn More
BINDER™ Vacuum Pumps VAP with Pump Module
SDP
Catalog No. 08111947 Shop All BINDER Inc. Products
Change view
Click to view available options
For Use With (Equipment):
Model VD 115
Type:
VAP 2
Catalog No. For Use With (Equipment) Type
08-111-947 Model VD 115 VAP 2
1 options
Catalog No. 08-111-947 Supplier BINDER™ Supplier No. 80121969
Combined set of vacuum pump and vacuum module including all connection parts.
  • Choice of chemistry diaphragm pumps that are optimally matched to the vacuum ovens.
  • Scoop is resistant to a variety of chemicals, especially acids
  • Final vacuum can be regulated via the vacuum oven
  • Pump module lifts the vacuum oven to the optimum working height, significantly reduces the noise of the vacuum pump, and protects the pump and glass material from mechanical damage.
  • Pump in the module can be switched on and off using a switchable socket via the control of the vacuum furnace.
Specifications
For Use With (Equipment) Model VD 115
Material Steel
Type VAP 2
Includes Pump module
Electrical Requirements 50/60 Hz
Voltage 120 V